Product Details
- SNP ID
-
rs116969157
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57416947 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTAGTGTCTCTTCCTCCTTCAGAA[G/T]TGGGTGGAGGCTAGCAGGGTGTGGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF17
PubMed Links
Gene Details
- Gene
- ZNF17
- Gene Name
- zinc finger protein 17
View Full Product Details