Product Details

SNP ID
rs112895297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTTTGCACTCAAAGGGCTTCT[A/C]TCCGGTATGGGTCCTCTTATGTCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 1646 Nonsense Mutation GAG,TAG E420* NP_001007249.1
XM_017026369.1 1646 Nonsense Mutation GAG,TAG E383* XP_016881858.1

View Full Product Details