Product Details

SNP ID
rs116196721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35140608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCTTTCCAGAAAGTCCAAAG[A/G]AACACGACCCGTTCACTTACGGTGA
Phenotype
MIM: 602359 MIM: 606684 MIM: 608303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXYD1 PubMed Links

Gene Details

Gene
FXYD1
Gene Name
FXYD domain containing ion transport regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278717.1 162 Missense Mutation AAA,GAA K25E NP_001265646.1
NM_001278718.1 162 Missense Mutation AAA,GAA K25E NP_001265647.1
NM_005031.4 162 Missense Mutation AAA,GAA K25E NP_005022.2
NM_021902.3 162 Missense Mutation AAA,GAA K25E NP_068702.1
XM_017026874.1 162 Missense Mutation AAA,GAA K25E XP_016882363.1
XM_017026875.1 162 Missense Mutation AAA,GAA K43E XP_016882364.1
XM_017026876.1 162 Missense Mutation AAA,GAA K43E XP_016882365.1
Gene
FXYD7
Gene Name
FXYD domain containing ion transport regulator 7
There are no transcripts associated with this gene.

Gene
LGI4
Gene Name
leucine rich repeat LGI family member 4
There are no transcripts associated with this gene.

View Full Product Details