Product Details
- SNP ID
-
rs117559137
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:36743806 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACACACCAGGTACAAGAGAAAA[A/C]CTCAATGTCGTAAAGATGTCAATAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF850
PubMed Links
Gene Details
- Gene
- ZNF850
- Gene Name
- zinc finger protein 850
View Full Product Details