Product Details

SNP ID
rs117193629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1104875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCGGCGCCGGCGGAAGAAGCC[C/T]TGTCCCCGCAGCTTGCGACCGGAGA
Phenotype
MIM: 138322 MIM: 180664 MIM: 615729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX4 PubMed Links

Gene Details

Gene
GPX4
Gene Name
glutathione peroxidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039847.2 226 Intron NP_001034936.1
NM_001039848.2 226 Silent Mutation CCC,CCT P47P NP_001034937.1
NM_002085.4 226 Intron NP_002076.2
Gene
POLR2E
Gene Name
RNA polymerase II subunit E
There are no transcripts associated with this gene.

Gene
SBNO2
Gene Name
strawberry notch homolog 2
There are no transcripts associated with this gene.

View Full Product Details