Product Details

SNP ID
rs116920201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2294993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACCAGGTTGTGCGTCTCCTCA[C/T]GGTTCCCACTGGCCTTCGATCCTCA
Phenotype
MIM: 609792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINGO3 PubMed Links

Gene Details

Gene
LINGO3
Gene Name
leucine rich repeat and Ig domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101391.1 Intron NP_001094861.1

View Full Product Details