Product Details

SNP ID
rs114622254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4280186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCGCGCGCAGAAGAACCTGGA[C/T]TTCGAGGACCCCTATGAGGACGCGG
Phenotype
MIM: 610481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHD PubMed Links

Gene Details

Gene
SHD
Gene Name
Src homology 2 domain containing transforming protein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020209.3 1586 Silent Mutation GAC,GAT D41D NP_064594.3

View Full Product Details