Product Details

SNP ID
rs112105291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4548077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGAGCCACAGGCACTCGGACCAC[A/G]CAGCGGGGGAAGGCAGCCAGCAGGC
Phenotype
MIM: 611289 MIM: 608873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRG1 PubMed Links

Gene Details

Gene
LRG1
Gene Name
leucine rich alpha-2-glycoprotein 1
There are no transcripts associated with this gene.

Gene
SEMA6B
Gene Name
semaphorin 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032108.3 1813 Silent Mutation NP_115484.2
XM_011527639.2 1813 Silent Mutation XP_011525941.1
XM_011527640.2 1813 Silent Mutation XP_011525942.1
XM_011527641.1 1813 Silent Mutation XP_011525943.1

View Full Product Details