Product Details
- SNP ID
-
rs116475299
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4771069 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGAGACCCAATGAGGGAATGACA[C/T]TTAAAGCCCTCCCTAGACCAGAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7-3
PubMed Links
Gene Details
- Gene
- MIR7-3
- Gene Name
- microRNA 7-3
There are no transcripts associated with this gene.
- Gene
- MIR7-3HG
- Gene Name
- MIR7-3 host gene
There are no transcripts associated with this gene.
View Full Product Details