Product Details

SNP ID
rs117320342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10416969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGGAGTCGCAGTGCCCTGTC[C/G]GTGGCAGGGACCGGGGACGAGGTGC
Phenotype
MIM: 600126
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDE4A PubMed Links

Gene Details

Gene
PDE4A
Gene Name
phosphodiesterase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111307.1 106 Intron NP_001104777.1
NM_001111308.1 106 Intron NP_001104778.1
NM_001111309.1 106 Intron NP_001104779.1
NM_001243121.1 106 Silent Mutation TCC,TCG S9S NP_001230050.1
NM_006202.2 106 Intron NP_006193.1
XM_005259933.2 106 Intron XP_005259990.1
XM_011528054.1 106 Silent Mutation TCC,TCG S9S XP_011526356.1
XM_011528055.2 106 Intron XP_011526357.1
XM_017026865.1 106 Intron XP_016882354.1

View Full Product Details