Product Details

SNP ID
rs111814574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11541198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGATGGCATCATTCTTTGCGA[A/G]TGAGTGAGGCTCTCGAAGCCGAGAC
Phenotype
MIM: 600806
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNN1 PubMed Links

Gene Details

Gene
CNN1
Gene Name
calponin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001299.5 Intron NP_001290.2
NM_001308341.1 Intron NP_001295270.1
NM_001308342.1 Intron NP_001295271.1
XM_005259741.2 Intron XP_005259798.1
XM_017026289.1 Intron XP_016881778.1

View Full Product Details