Product Details

SNP ID
rs113108880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14546074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTGGGGGACCTGCCTGGAAGG[G/T]GAGGAAGTGATCCATGCTAGCTCGT
Phenotype
MIM: 610057
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TECR PubMed Links

Gene Details

Gene
TECR
Gene Name
trans-2,3-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321170.1 Intron NP_001308099.1
NM_138501.5 Intron NP_612510.1
XM_006722945.1 Intron XP_006723008.1
XM_011528442.2 Intron XP_011526744.1

View Full Product Details