Product Details

SNP ID
rs115141412
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17240583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTGAAAGGGAGGGGTGAAAGG[A/G]AGGGGAAAAAGGGGAGGAAGAAGCT
Phenotype
MIM: 132880 MIM: 611810
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links
Additional Information
For this assay, SNP(s) [rs201503736] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005234.3 Intron NP_005225.2
Gene
USHBP1
Gene Name
USH1 protein network component harmonin binding protein 1
There are no transcripts associated with this gene.

View Full Product Details