Product Details

SNP ID
rs114221795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43458084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTACAACGTCCGGGAGAAGGA[C/T]ATCCAGCGCTTTTTCAGTGGCTATG
Phenotype
MIM: 601944
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF6 PubMed Links

Gene Details

Gene
SRSF6
Gene Name
serine and arginine rich splicing factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006275.5 221 Silent Mutation GAC,GAT D17D NP_006266.2

View Full Product Details