Product Details

SNP ID
rs111774248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45796672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATAGGGAGACATCAGGAAAGGT[C/T]GCCAATAGTCGCTGGCTCACTTTCT
Phenotype
MIM: 611388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP1 PubMed Links

Gene Details

Gene
DNTTIP1
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052951.2 Intron NP_443183.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
There are no transcripts associated with this gene.

View Full Product Details