Product Details
- SNP ID
-
rs111856613
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45824640 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGTCAAAGGCAGCCTTGAACTC[A/T]GCGAGGGAGGGCAGAGGGCAGAGGT
- Phenotype
-
MIM: 191039
MIM: 605574
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNX21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs556074291] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNX21
- Gene Name
- sorting nexin family member 21
There are no transcripts associated with this gene.
- Gene
- TNNC2
- Gene Name
- troponin C2, fast skeletal type
- Gene
- UBE2C
- Gene Name
- ubiquitin conjugating enzyme E2 C
There are no transcripts associated with this gene.
View Full Product Details