Product Details

SNP ID
rs113214229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50739474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAAAGAAAGTAGCTTGAGAGA[A/G]TGGGTGGGGATTATCTTTGAAGGTG
Phenotype
MIM: 608975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARD6B PubMed Links

Gene Details

Gene
PARD6B
Gene Name
par-6 family cell polarity regulator beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032521.2 Intron NP_115910.1

View Full Product Details