Product Details

SNP ID
rs113751177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31423314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAGAAGGGGGTGAGGTGAAAG[C/T]AGAAATGCTGAAAGACTGAACATTT
Phenotype
MIM: 616075 MIM: 616077
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DEFB121 PubMed Links

Gene Details

Gene
DEFB121
Gene Name
defensin beta 121
There are no transcripts associated with this gene.

Gene
DEFB122
Gene Name
defensin beta 122 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details