Product Details

SNP ID
rs117088963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1631595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCCTGTATCCATCTCTAGTCTG[C/T]CTTACAGAGAAACTGACCTTGGTAA
Phenotype
MIM: 605466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIRPG PubMed Links

Gene Details

Gene
SIRPG
Gene Name
signal regulatory protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039508.1 Intron NP_001034597.1
NM_018556.3 Intron NP_061026.2
NM_080816.2 Intron NP_543006.2
XM_005260749.3 Intron XP_005260806.1
XM_011529286.2 Intron XP_011527588.1
XM_011529287.2 Intron XP_011527589.1
Gene
SIRPG-AS1
Gene Name
SIRPG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details