Product Details

SNP ID
rs112136447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:21395970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACGCCGCCGCTGTACTCCCCGG[C/G]GGCCGCGTCCAGGGCCGCCAGGCCG
Phenotype
MIM: 607808 MIM: 608851
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NKX2-4 PubMed Links

Gene Details

Gene
NKX2-4
Gene Name
NK2 homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033176.1 1016 Missense Mutation CCC,GCC P336A NP_149416.1
Gene
XRN2
Gene Name
5'-3' exoribonuclease 2
There are no transcripts associated with this gene.

View Full Product Details