Product Details

SNP ID
rs114937908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23605620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGGTCACCAACCTTTCTGTCC[A/C]TGCTATCCTCCCTCCATGAACTCAG
Phenotype
MIM: 616543
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CST9 PubMed Links

Gene Details

Gene
CST9
Gene Name
cystatin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008693.2 354 Missense Mutation AGG,ATG R82M NP_001008693.2

View Full Product Details