Product Details

SNP ID
rs121909394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3228337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGCTCATGCCGAAGGCACAC[G/A]GCAGCAGCAGCTGAAGCACCTGCAG
Phenotype
MIM: 147520 MIM: 610206
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ITPA PubMed Links

Gene Details

Gene
ITPA
Gene Name
inosine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267623.1 2603 Intron NP_001254552.1
NM_001324236.1 2603 Intron NP_001311165.1
NM_001324237.1 2603 Intron NP_001311166.1
NM_001324238.1 2603 Intron NP_001311167.1
NM_001324240.1 2603 Intron NP_001311169.1
NM_033453.3 2603 Intron NP_258412.1
NM_181493.3 2603 Intron NP_852470.1
XM_006723564.3 2603 Intron XP_006723627.1
XM_006723565.3 2603 Intron XP_006723628.1
XM_011529234.2 2603 Intron XP_011527536.1
Gene
SLC4A11
Gene Name
solute carrier family 4 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174089.1 2603 Missense Mutation CCG,CTG P827L NP_001167560.1
NM_001174090.1 2603 Missense Mutation CCG,CTG P870L NP_001167561.1
NM_032034.3 2603 Missense Mutation CCG,CTG P843L NP_114423.1
XM_005260856.4 2603 Missense Mutation CCG,CTG P950L XP_005260913.1
XM_005260857.1 2603 Missense Mutation CCG,CTG P808L XP_005260914.1
XM_011529383.2 2603 Missense Mutation CCG,CTG P816L XP_011527685.1
XM_011529384.1 2603 Missense Mutation CCG,CTG P808L XP_011527686.1
XM_011529385.1 2603 Missense Mutation CCG,CTG P808L XP_011527687.1
XM_017028093.1 2603 Missense Mutation CCG,CTG P948L XP_016883582.1
XM_017028094.1 2603 Missense Mutation CCG,CTG P808L XP_016883583.1
XM_017028095.1 2603 Missense Mutation CCG,CTG P789L XP_016883584.1
XM_017028096.1 2603 Missense Mutation CCG,CTG P808L XP_016883585.1
XM_017028097.1 2603 Intron XP_016883586.1

View Full Product Details