Product Details

SNP ID
rs111402070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42405896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGTGTGGGAGGCGAGCCTGGG[A/G]ATCAGGGTCGCTGATGCTGGGGTGA
Phenotype
MIM: 605511 MIM: 605736
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS3 PubMed Links

Gene Details

Gene
TMPRSS3
Gene Name
transmembrane protease, serine 3
There are no transcripts associated with this gene.

Gene
UBASH3A
Gene Name
ubiquitin associated and SH3 domain containing A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001895.2 Intron NP_001001895.1
NM_001243467.1 Intron NP_001230396.1
NM_018961.3 Intron NP_061834.1
XM_006724013.3 Intron XP_006724076.1
XM_011529605.2 Intron XP_011527907.1
XM_011529606.2 Intron XP_011527908.1
XM_011529607.2 Intron XP_011527909.1
XM_011529609.1 Intron XP_011527911.1
XM_011529610.2 Intron XP_011527912.1

View Full Product Details