Product Details

SNP ID
rs117906779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31219904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCTTAACAATCAGTAGCCACT[A/G]TTGATAAGTCTGCAAATATGAGAGG
Phenotype
MIM: 601988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMK2 PubMed Links

Gene Details

Gene
LIMK2
Gene Name
LIM domain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031801.1 Intron NP_001026971.1
NM_005569.3 Intron NP_005560.1
NM_016733.2 Intron NP_057952.1

View Full Product Details