Product Details

SNP ID
rs111634119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31683398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGTAGGAGCTCCTTAAACAT[A/G]CCTGGACAAGGGCAAAATGTACTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRR14L PubMed Links

Gene Details

Gene
PRR14L
Gene Name
proline rich 14 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173566.2 9338 UTR 3 NP_775837.2
XM_006724219.3 9338 UTR 3 XP_006724282.1
XM_006724220.3 9338 UTR 3 XP_006724283.1
XM_006724221.2 9338 UTR 3 XP_006724284.1
XM_011530080.2 9338 UTR 3 XP_011528382.1
XM_011530081.2 9338 UTR 3 XP_011528383.1
XM_017028736.1 9338 UTR 3 XP_016884225.1
XM_017028737.1 9338 UTR 3 XP_016884226.1

View Full Product Details