Product Details

SNP ID
rs115833761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21634014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCCCCCTGTGACATACCTGT[C/T]TGCTCAGGTGCAGCTGCCCAAGAAG
Phenotype
MIM: 607551 MIM: 603721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC116 PubMed Links

Gene Details

Gene
CCDC116
Gene Name
coiled-coil domain containing 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152612.2 Intron NP_689825.2
XM_006724159.2 Intron XP_006724222.1
XM_011529984.2 Intron XP_011528286.1
XM_011529985.1 Intron XP_011528287.1
Gene
SDF2L1
Gene Name
stromal cell derived factor 2 like 1
There are no transcripts associated with this gene.

Gene
UBE2L3
Gene Name
ubiquitin conjugating enzyme E2 L3
There are no transcripts associated with this gene.

Gene
YDJC
Gene Name
YdjC homolog (bacterial)
There are no transcripts associated with this gene.

View Full Product Details