Product Details

SNP ID
rs113511992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35709126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGTGGTGGACTTGAAATGAC[C/T]TGTGTTCACTTCCTTTTACTTATCG
Phenotype
MIM: 607255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOL5 PubMed Links

Gene Details

Gene
APOL5
Gene Name
apolipoprotein L5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030642.1 Intron NP_085145.1
XM_006724321.3 Intron XP_006724384.1
XM_017028945.1 Intron XP_016884434.1

View Full Product Details