Product Details

SNP ID
rs111523609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39501433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGTTAGGAAACTTGGCAGAG[G/T]GCAGCTGTCCCTAAGTGGTGGAACT
Phenotype
MIM: 604621 MIM: 615497
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MGAT3 PubMed Links

Gene Details

Gene
MGAT3
Gene Name
mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.

Gene
MIEF1
Gene Name
mitochondrial elongation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304564.1 Intron NP_001291493.1
NM_019008.5 Intron NP_061881.2
XM_011530236.1 Intron XP_011528538.1
XM_011530237.1 Intron XP_011528539.1
XM_011530238.1 Intron XP_011528540.1
XM_017028838.1 Intron XP_016884327.1

View Full Product Details