Product Details

SNP ID
rs115659581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46277255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACCCCGAGAAACCACAGGAAAG[A/G]AAACTTGTTTCATGTGTGACCACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC38 PubMed Links

Gene Details

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 Intron NP_060401.2
XM_011530259.1 Intron XP_011528561.1
XM_011530260.2 Intron XP_011528562.1

View Full Product Details