Product Details

SNP ID
rs111837673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27980305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATTACCATTTTTTCCCCTACT[A/G]ACATAAAGAAACCCTCATAATTGCT
Phenotype
MIM: 615098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC28 PubMed Links

Gene Details

Gene
TTC28
Gene Name
tetratricopeptide repeat domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145418.1 9504 UTR 3 NP_001138890.1
XM_005261405.2 9504 UTR 3 XP_005261462.1
XM_006724171.3 9504 UTR 3 XP_006724234.1
XM_011530018.2 9504 UTR 3 XP_011528320.1
XM_011530019.2 9504 Intron XP_011528321.1
XM_011530020.1 9504 Intron XP_011528322.1
XM_011530021.2 9504 Intron XP_011528323.1
XM_011530022.1 9504 Intron XP_011528324.1
XM_017028673.1 9504 UTR 3 XP_016884162.1
Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details