Product Details

SNP ID
rs115209402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54533117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAGGAAACTGAAGCTTAGCAA[A/G]GTTAAGTGACTTGATCTGAGGTGAA
Phenotype
MIM: 300873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNL3L PubMed Links

Gene Details

Gene
GNL3L
Gene Name
G protein nucleolar 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184819.1 Intron NP_001171748.1
NM_019067.5 Intron NP_061940.1

View Full Product Details