Product Details

SNP ID
rs116637437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:145829407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTTTTGCCAAGGCTGCTTGTG[A/C]TACCAGTTTGCTGGTGGCTAACGGC
Phenotype
MIM: 300561 MIM: 300565
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLITRK2 PubMed Links

Gene Details

Gene
SLITRK2
Gene Name
SLIT and NTRK like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144003.2 1998 Intron NP_001137475.1
NM_001144004.2 1998 Intron NP_001137476.1
NM_001144005.2 1998 Intron NP_001137477.1
NM_001144006.2 1998 Intron NP_001137478.1
NM_001144008.2 1998 Intron NP_001137480.1
NM_001144009.2 1998 Intron NP_001137481.1
NM_001144010.2 1998 Intron NP_001137482.1
NM_032539.4 1998 Intron NP_115928.1
XM_005262342.3 1998 UTR 3 XP_005262399.1
XM_005262343.3 1998 UTR 3 XP_005262400.1
XM_005262344.3 1998 UTR 3 XP_005262401.1
XM_005262345.3 1998 UTR 3 XP_005262402.1
Gene
TMEM257
Gene Name
transmembrane protein 257
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004709.2 1998 UTR 3 NP_004700.1

View Full Product Details