Product Details

SNP ID
rs114494889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76162161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAATTAACATTTCCATGAGGTTT[C/T]ATTTGTGCTTAAGACAATTTCAATG
Phenotype
MIM: 602257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARB2 PubMed Links

Gene Details

Gene
SCARB2
Gene Name
scavenger receptor class B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204255.1 Intron NP_001191184.1
NM_005506.3 Intron NP_005497.1

View Full Product Details