Product Details

SNP ID
rs116121278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109710951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATATCTGTCCCTGGAAGAACTA[C/T]GTCAAAAGATTTTAAAGAGACAGGC
Phenotype
MIM: 601532 MIM: 611652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP6 PubMed Links

Gene Details

Gene
CASP6
Gene Name
caspase 6
There are no transcripts associated with this gene.

Gene
PLA2G12A
Gene Name
phospholipase A2 group XIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030821.4 4273 UTR 3 NP_110448.2

View Full Product Details