Product Details

SNP ID
rs112235071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75515860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGTTTACAATCTGAATGAAGGA[A/G]CTTTGCTCGAATATATTGCCCTCTC
Phenotype
MIM: 607680 MIM: 612535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCHY1 PubMed Links

Gene Details

Gene
RCHY1
Gene Name
ring finger and CHY zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009922.2 Intron NP_001009922.1
NM_001278536.1 Intron NP_001265465.1
NM_001278537.1 Intron NP_001265466.1
NM_001278538.1 Intron NP_001265467.1
NM_001278539.1 Intron NP_001265468.1
NM_015436.3 Intron NP_056251.2
XM_011531838.2 Intron XP_011530140.1
XM_011531839.2 Intron XP_011530141.1
Gene
THAP6
Gene Name
THAP domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317791.1 Intron NP_001304720.1
NM_144721.5 Intron NP_653322.1
XM_005262772.2 Intron XP_005262829.1
XM_005262774.3 Intron XP_005262831.1
XM_006714109.3 Intron XP_006714172.1
XM_011531666.2 Intron XP_011529968.1
XM_011531667.2 Intron XP_011529969.1
XM_017007800.1 Intron XP_016863289.1
XM_017007801.1 Intron XP_016863290.1

View Full Product Details