Product Details

SNP ID
rs116159892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:120218794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCAGGATATTCGTAAAGCACA[C/T]GGGAAACAAATCATGGATGATCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM185B PubMed Links

Gene Details

Gene
TMEM185B
Gene Name
transmembrane protein 185B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024121.2 4615 UTR 3 NP_077026.2

View Full Product Details