Product Details

SNP ID
rs114710451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:147930605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTTACCTCTCAGAATATTGGT[A/T]AGCAGTTATTTTCGCTTTACTCTGT
Phenotype
MIM: 102581 MIM: 603056
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACVR2A PubMed Links

Gene Details

Gene
ACVR2A
Gene Name
activin A receptor type 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278579.1 6495 UTR 3 NP_001265508.1
NM_001278580.1 6495 UTR 3 NP_001265509.1
NM_001616.4 6495 UTR 3 NP_001607.1
Gene
ORC4
Gene Name
origin recognition complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190879.2 6495 UTR 3 NP_001177808.1
NM_001190881.2 6495 Intron NP_001177810.1
NM_001190882.2 6495 UTR 3 NP_001177811.1
NM_002552.4 6495 UTR 3 NP_002543.2
NM_181741.3 6495 UTR 3 NP_859525.1
NM_181742.3 6495 Intron NP_859526.1
XM_011511255.2 6495 Intron XP_011509557.1
XM_017004226.1 6495 Intron XP_016859715.1
XM_017004227.1 6495 Intron XP_016859716.1

View Full Product Details