Product Details

SNP ID
rs118161769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95165624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCACAACGGCTCGCTCTCCCAG[C/G]TAAGGCGACAGCGCTGCCATTTTGG
Phenotype
MIM: 194500
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF2 PubMed Links

Gene Details

Gene
ZNF2
Gene Name
zinc finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017396.2 208 UTR 5 NP_001017396.1
NM_001282398.1 208 UTR 5 NP_001269327.1
NM_001291604.1 208 UTR 5 NP_001278533.1
NM_001291605.1 208 UTR 5 NP_001278534.1
NM_021088.3 208 UTR 5 NP_066574.2
Gene
ZNF514
Gene Name
zinc finger protein 514
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318005.1 208 Intron NP_001304934.1
NM_032788.2 208 Intron NP_116177.1
XM_006712806.2 208 Intron XP_006712869.1

View Full Product Details