Product Details

SNP ID
rs117462563
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95171890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAGGATCACCCCAGGTTCAGG[A/G]ATTCACTAGAAGGACTCAAGACTCG
Phenotype
MIM: 194500
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF2 PubMed Links

Gene Details

Gene
ZNF2
Gene Name
zinc finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017396.2 Intron NP_001017396.1
NM_001282398.1 Intron NP_001269327.1
NM_001291604.1 Intron NP_001278533.1
NM_001291605.1 Intron NP_001278534.1
NM_021088.3 Intron NP_066574.2
Gene
ZNF514
Gene Name
zinc finger protein 514
There are no transcripts associated with this gene.

View Full Product Details