Product Details

SNP ID
rs111952222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102358018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAATTATATTAAAAAAAAACTG[G/T]TGTTATGATTTTCATCCAGGCATTT
Phenotype
MIM: 604494 MIM: 601203
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL18R1 PubMed Links

Gene Details

Gene
IL18R1
Gene Name
interleukin 18 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282399.1 Intron NP_001269328.1
NM_003855.3 Intron NP_003846.1
XM_005264039.3 Intron XP_005264096.1
XM_005264040.3 Intron XP_005264097.1
XM_011512099.1 Intron XP_011510401.1
XM_017005181.1 Intron XP_016860670.1
XM_017005182.1 Intron XP_016860671.1
XM_017005183.1 Intron XP_016860672.1
XM_017005184.1 Intron XP_016860673.1
Gene
IL1RL1
Gene Name
interleukin 1 receptor like 1
There are no transcripts associated with this gene.

View Full Product Details