Product Details

SNP ID
rs117803955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31335388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGCAGCTTCACAAAATCAGAT[C/G]CAATTAACCTTGAATTTGTTTCATT
Phenotype
MIM: 607633
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XDH PubMed Links

Gene Details

Gene
XDH
Gene Name
xanthine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000379.3 4651 UTR 3 NP_000370.2
XM_011533095.2 4651 UTR 3 XP_011531397.1
XM_011533096.2 4651 Intron XP_011531398.1

View Full Product Details