Product Details

SNP ID
rs114849961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48537484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTATATTATTCTGTAGGGTAT[C/T]GGTTGCTAGTATTTTACTTTGGATT
Phenotype
MIM: 605357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STON1 PubMed Links

Gene Details

Gene
STON1
Gene Name
stonin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198595.1 Intron NP_001185524.1
NM_006873.3 Intron NP_006864.2
Gene
STON1-GTF2A1L
Gene Name
STON1-GTF2A1L readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198593.1 Intron NP_001185522.1
NM_001198594.1 Intron NP_001185523.1
NM_172311.2 Intron NP_758515.1

View Full Product Details