Product Details

SNP ID
rs117628715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71187078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGTAGCAATCAGAAAGAAGCC[A/G]CTATTATCTTCAGGGCTCTCCAATT
Phenotype
MIM: 611018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAIP2B PubMed Links

Gene Details

Gene
PAIP2B
Gene Name
poly(A) binding protein interacting protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020459.1 1930 UTR 3 NP_065192.1
XM_005264310.3 1930 UTR 3 XP_005264367.1
XM_005264311.4 1930 UTR 3 XP_005264368.1
XM_011532842.2 1930 UTR 3 XP_011531144.1
XM_011532843.2 1930 UTR 3 XP_011531145.1

View Full Product Details