Product Details

SNP ID
rs113692426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28829223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCATGGGCTTTAGGGAAAAA[C/T]TGGAGAAAATTGTTCCCTAATTTCT
Phenotype
MIM: 614029
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPDYA PubMed Links

Gene Details

Gene
SPDYA
Gene Name
speedy/RINGO cell cycle regulator family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008779.1 662 Silent Mutation AAC,AAT N152N NP_001008779.1
NM_001142634.1 662 Silent Mutation AAC,AAT N152N NP_001136106.1
NM_182756.3 662 Silent Mutation AAC,AAT N152N NP_877433.2

View Full Product Details