Product Details

SNP ID
rs115958679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:88153426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCACAACTGAAATTGAAGGAAAC[A/G]CCAGAGCCAGTTACAAAGACAGAGG
Phenotype
MIM: 603363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf38 PubMed Links

Gene Details

Gene
C3orf38
Gene Name
chromosome 3 open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173824.3 670 Silent Mutation ACA,ACG T110T NP_776185.2
XM_005264745.4 670 Silent Mutation ACA,ACG T110T XP_005264802.1
Gene
CGGBP1
Gene Name
CGG triplet repeat binding protein 1
There are no transcripts associated with this gene.

Gene
ZNF654
Gene Name
zinc finger protein 654
There are no transcripts associated with this gene.

View Full Product Details