Product Details

SNP ID
rs114364578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23923551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTTTTGGTATAACTGCATGGT[A/G]TCCCATCATGTGGATGTACCATGAT
Phenotype
MIM: 604496 MIM: 604174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
There are no transcripts associated with this gene.

Gene
RPL15
Gene Name
ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253379.1 2694 Intron NP_001240308.1
NM_001253380.1 2694 Intron NP_001240309.1
NM_001253382.1 2694 Intron NP_001240311.1
NM_001253383.1 2694 Intron NP_001240312.1
NM_001253384.1 2694 UTR 3 NP_001240313.1
NM_002948.3 2694 Intron NP_002939.2

View Full Product Details