Product Details

SNP ID
rs114140352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33502122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGTCAGCACTCCTCACTATC[C/T]GATAAAACCTAAATTCATTGGTTTG
Phenotype
MIM: 605853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLASP2 PubMed Links

Gene Details

Gene
CLASP2
Gene Name
cytoplasmic linker associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207044.1 Intron NP_001193973.1
NM_015097.2 Intron NP_055912.2
XM_006713040.1 Intron XP_006713103.1
XM_006713041.1 Intron XP_006713104.1
XM_006713042.1 Intron XP_006713105.1
XM_006713048.1 Intron XP_006713111.1
XM_006713049.1 Intron XP_006713112.1
XM_006713050.1 Intron XP_006713113.1
XM_006713052.1 Intron XP_006713115.1
XM_006713053.1 Intron XP_006713116.1
XM_011533515.1 Intron XP_011531817.1
XM_017005947.1 Intron XP_016861436.1
XM_017005948.1 Intron XP_016861437.1
XM_017005949.1 Intron XP_016861438.1
XM_017005950.1 Intron XP_016861439.1
XM_017005951.1 Intron XP_016861440.1
XM_017005952.1 Intron XP_016861441.1
XM_017005953.1 Intron XP_016861442.1
XM_017005954.1 Intron XP_016861443.1
XM_017005955.1 Intron XP_016861444.1
XM_017005956.1 Intron XP_016861445.1
XM_017005957.1 Intron XP_016861446.1
XM_017005958.1 Intron XP_016861447.1
XM_017005959.1 Intron XP_016861448.1
XM_017005960.1 Intron XP_016861449.1
XM_017005961.1 Intron XP_016861450.1
XM_017005962.1 Intron XP_016861451.1
XM_017005963.1 Intron XP_016861452.1
XM_017005964.1 Intron XP_016861453.1
XM_017005965.1 Intron XP_016861454.1
XM_017005966.1 Intron XP_016861455.1
XM_017005967.1 Intron XP_016861456.1
XM_017005968.1 Intron XP_016861457.1
XM_017005969.1 Intron XP_016861458.1
XM_017005970.1 Intron XP_016861459.1
XM_017005971.1 Intron XP_016861460.1
XM_017005972.1 Intron XP_016861461.1
XM_017005973.1 Intron XP_016861462.1
XM_017005974.1 Intron XP_016861463.1
XM_017005975.1 Intron XP_016861464.1
XM_017005976.1 Intron XP_016861465.1
XM_017005977.1 Intron XP_016861466.1
XM_017005978.1 Intron XP_016861467.1
XM_017005979.1 Intron XP_016861468.1
XM_017005980.1 Intron XP_016861469.1
XM_017005981.1 Intron XP_016861470.1
XM_017005982.1 Intron XP_016861471.1
XM_017005983.1 Intron XP_016861472.1
XM_017005984.1 Intron XP_016861473.1
XM_017005985.1 Intron XP_016861474.1
XM_017005986.1 Intron XP_016861475.1

View Full Product Details