Product Details

SNP ID
rs111543377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40309706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGTCTCCATCGGCCATTTTGTG[C/G]GAGAAGCCGCAGCGCCGCCTCTTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF1B PubMed Links

Gene Details

Gene
EIF1B
Gene Name
eukaryotic translation initiation factor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005875.2 25 UTR 5 NP_005866.1
Gene
EIF1B-AS1
Gene Name
EIF1B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details