Product Details

SNP ID
rs116432956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44440008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCAGATCCAGGCAAACTGAGG[A/G]GGAGAGCTTGAATGCTATTAGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF445 PubMed Links

Gene Details

Gene
ZNF445
Gene Name
zinc finger protein 445
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181489.5 9105 UTR 3 NP_852466.1
XM_005265102.2 9105 UTR 3 XP_005265159.1
XM_005265103.3 9105 UTR 3 XP_005265160.1
XM_011533674.2 9105 UTR 3 XP_011531976.1
XM_011533675.2 9105 UTR 3 XP_011531977.1
XM_017006345.1 9105 UTR 3 XP_016861834.1
XM_017006346.1 9105 UTR 3 XP_016861835.1

View Full Product Details