Product Details

SNP ID
rs115077448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48469572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGATGTGGTGGTGGTGACAAC[C/T]GCTGCAAAGAGAATTCCAGTCAGGA
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272065.1 748 Intron NP_001258994.1
NM_001272066.1 748 Silent Mutation CCA,CCG P113P NP_001258995.1
NM_001272067.1 748 Silent Mutation CCA,CCG P113P NP_001258996.1
NM_001272068.1 748 Silent Mutation CCA,CCG P113P NP_001258997.1
NM_001272082.1 748 Silent Mutation CCA,CCG P41P NP_001259011.1
NM_001272083.1 748 Silent Mutation CCA,CCG P41P NP_001259012.1
NM_016479.4 748 Intron NP_057563.3
XM_005265205.1 748 Silent Mutation CCA,CCG P113P XP_005265262.1
XM_006713188.2 748 Silent Mutation CCA,CCG P113P XP_006713251.1
XM_011533796.1 748 Silent Mutation CCA,CCG P113P XP_011532098.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
There are no transcripts associated with this gene.

View Full Product Details